Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene glycol BioUlt1KG
Catalog No. 502898457
Encompass
Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system which is required for the purification of biological materials. PEG also interacts with cell membrane thereby allowing cell fusion.
Catalog No. 50-289-8457 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 812551KG
Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system which is required for the purification of biological materials. PEG also interacts with cell membrane thereby allowing cell fusion.