Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Bis(3-aminopropyl) Ether, TCI America™
SDP

Catalog No. P19481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19481G Supplier TCI America Supplier No. P19481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34901-14-9
MDL Number MFCD00197739

Specifications

Melting Point 54°C
Quantity 1 g
Physical Form Crystalline Lumps
Chemical Name or Material Polyethylene Glycol Bis(3-aminopropyl) Ether
EINECSNumber (7)-1324
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.