Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Dimethyl Ether (Mw.=ca. 240), TCI America™
SDP

Catalog No. P224125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P224125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. P224125G Supplier TCI America Supplier No. P224125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 24991-55-7
MDL Number MFCD00147972

Specifications

Color Yellow
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Polyethylene Glycol Dimethyl Ether (Mw.=ca. 240)
EINECSNumber (7)-1321
RTECSNumber MC9630000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.