Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Distearate, MP Biomedicals

Catalog No. ICN217565 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217565 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217565 Supplier MP Biomedicals, Inc Supplier No. 0521756525

Chemical Identifiers

CAS 9005-08-7
Molecular Formula C44H86O7
Molecular Weight (g/mol) 727.17
InChI Key MQFYRUGXOJAUQK-UHFFFAOYSA-N
Synonym ethylene glycol; heptadecanoic acid, peg 400 distearate, peg 600 distearate, peg 4000 distearate, peg 6000 distearate, poly ethylene glycol distearate, polyethylene glycol 400 distearate, poly ethylene glycol n distearate, poly oxy-1,2-ethanediyl , ?-1-oxooctadecyl-?-1-oxooctadecyl oxy
PubChem CID 16211945
IUPAC Name ethane-1,2-diol;heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O.C(CO)O

Specifications

Quantity 25 g
Chemical Name or Material Polyethylene Glycol Distearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.