Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene glycol for mo
Catalog No. 502891459
Encompass
Polyethylene glycol (PEG) is used in detergents, emulsifying agents, soaps, metal costing process, pharmaceutical preparations and cosmetics. PEG is commonly used as a lubricant. It application is also observed in chromatographic stationary phases. PEG prevents crystal formation in emulsions.
Catalog No. 50-289-1459 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P54132KG
Polyethylene glycol (PEG) is used in detergents, emulsifying agents, soaps, metal costing process, pharmaceutical preparations and cosmetics. PEG is commonly used as a lubricant. It application is also observed in chromatographic stationary phases. PEG prevents crystal formation in emulsions.