Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Glycidyl Lauryl Ether, TCI America™
SDP

Catalog No. P229425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P229425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P229425G Supplier TCI America Supplier No. P229425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86630-59-3
Molecular Formula C17H34O3
Molecular Weight (g/mol) 286.456
InChI Key ZCCBLVOOWREKMV-UHFFFAOYSA-N
PubChem CID 22986560
IUPAC Name 2-(2-dodecoxyethoxymethyl)oxirane
SMILES CCCCCCCCCCCCOCCOCC1CO1

Specifications

Color White
Quantity 25 g
Physical Form Paste
Chemical Name or Material Polyethylene Glycol Glycidyl Lauryl Ether
EINECSNumber (7)-1319&(7)-1281
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.