Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Monolaurate (n=approx. 10), TCI America™
SDP

Catalog No. P087225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P087225G Supplier TCI America Supplier No. P087225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31943-11-0
MDL Number MFCD00148005
Synonym Polyethylene Glycol Monododecanoate

Specifications

Color Yellow
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Polyethylene Glycol Monolaurate (n=approx. 10)
EINECSNumber (7)-0088
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.