Learn More
Medchemexpress LLC Polyethylene glycol monooleyl ether | 9004-98-2 | MFCD00074369 | 98.0% | 709 g/mol | (C2H4O)xC18H36O | 5 G
SDP
Catalog No. 5000282107
Encompass_Preferred
Polyoxyethylene(10) oleyl ether is a nonionic surfactant used in laboratory research to solubilize and emulsify hydrophobic compounds for aqueous formulations and sample preparation. It offers predictable ethylene oxide chain length and consistent performance in formulation workflows.
  • Nonionic surfactant for emulsification and solubilization.
  • High purity: 98.0%.
  • Average molecular weight approximately 709 g/mol.
  • Phase-change behavior: solid below 25 °C and liquid above 30 °C.
  • Suitable for formulation development and reagent preparation.
  • Available in small quantities for bench-scale experiments.
Catalog No. 50-002-82107 Supplier Medchemexpress LLC Supplier No. HYW2501685G
Polyoxyethylene(10) oleyl ether is a nonionic surfactant used in laboratory research to solubilize and emulsify hydrophobic compounds for aqueous formulations and sample preparation. It offers predictable ethylene oxide chain length and consistent performance in formulation workflows.
  • Nonionic surfactant for emulsification and solubilization.
  • High purity: 98.0%.
  • Average molecular weight approximately 709 g/mol.
  • Phase-change behavior: solid below 25 °C and liquid above 30 °C.
  • Suitable for formulation development and reagent preparation.
  • Available in small quantities for bench-scale experiments.