Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol, MP Biomedicals™

Catalog No. MP021954455 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021954455 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021954455 Supplier MP Biomedicals, Inc Supplier No. 02195445.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used to precipitate DNA. Polyethylene Glycol, MP Biomedicals (PEG) is a condensation polymer of ethylene oxide and water. PEGs are susceptible to oxidative degradation in the presence of air. PEGs do not hydrolyze or deteriorate upon storage. PEGs

PEG has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules, and in induction of cell hybridization. PEG 8000 can be used to precipitate DNA. PEG has also been used to recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples. PEG has demonstrated to have anti-cell proliferation and antitumor effects via induction of apoptosis.

  • PEGs are susceptible to oxidative degradation in the presence of air
    • Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant, can limit the amount of degradation
  • PEGs do not hydrolyze or deteriorate upon storage
  • PEGs do not support the growth of molds
  • PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym Carbowax, PEG, Poly(Ethylene Oxide), Alkapol PEG-200
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Melting Point 56°C to 66°C
Density 1.1-1.2g/mL (Lit.)
pH 5.0 to 7.0 (5% aq. soln.)
Quantity 500 g
Solubility Information Soluble in aqueous solution (50mg/mL, clear, colorless solution).
Vapor Pressure <0.01mmHg at 20°C (Lit.)
Formula Weight ∼8000
Physical Form Powder
Recommended Storage Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.