Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol, MP Biomedicals™

Catalog No. MP21948395 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21948395 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21948395 Supplier MP Biomedicals, Inc Supplier No. 02194839.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Also known as PEG. Polyethylene Glycol, MP Biomedicals is a condensation polymer of ethylene oxide and water. It is used in precipitation of proteins, separating and purifying biomolecules, induction of cell hybridization, and as a fusing agent in enhanci

PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym Carbowax, PEG, Poly(Ethylene Oxide), Alkapol PEG-8000, Macrogol, α-Hydro-o-hydroxypoly (oxy-1,2-ethanediyl)
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form White Prills
Quantity 500 g
Solubility Information Soluble in water (approximately 630mg/mL, 20°C). PEGs are also soluble in many polar solvents such as acetone, alcohols and chlorinated solvents. They are insoluble in nonpolar solvents such as hydrocarbons.
Formula Weight 8000g/mol
Grade Molecular Biology
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.