Learn More
Kyfora Bio Polyethylene glycol MW 600
Catalog No. 5000285307
Encompass
Polyethylene glycol (PEG) is a water-soluble biologically inert polymer that has a number of biological and pharmaceutical applications Specifically PEG600 can be used as an excipient in pharmaceutical products or as a coating on nanoparticles to prevent opsonization in vivo It can also be used for purification of plasmid DNA The polymer chains are hydroxyl terminated at both ends
Catalog No. 50-002-85307 Supplier Kyfora Bio Supplier No. 00684250
Polyethylene glycol (PEG) is a water-soluble biologically inert polymer that has a number of biological and pharmaceutical applications Specifically PEG600 can be used as an excipient in pharmaceutical products or as a coating on nanoparticles to prevent opsonization in vivo It can also be used for purification of plasmid DNA The polymer chains are hydroxyl terminated at both ends