Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polyethylene Glycol Standards Kit, 10 GPC Standards, Aqueous, Solid Formulation

Catalog No. 50785373 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit provides a unique collection of 10 GPC standards for aqueous molecular weight determination.

  • Kit contains 10 GPC standards.
  • Designed for aqueous molecular weight determination.
  • Includes 10 vials, each with 1.0 g of molecular weight standards.
  • Molecular weight standards included: 100, 200, 400, 600, 1000, 1500, 4300, 7000, 13000, 22000.
  • Separation mode: GPC/GFC/SEC.
  • Detection methods: UV, RI/ELSD.
  • Formulation: Solid.
  • Storage condition: 4°C.
  • System type: LC.
  • Shelf life of 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient.
  • Tolerance: 10% (Mp).
  • Type: Calibration.
  • Application: Gel filtration.

Catalog No. 50-785-373 Supplier Waters Corp Supplier No. WAT035711
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit provides a unique collection of 10 GPC standards for aqueous molecular weight determination.

  • Kit contains 10 GPC standards.
  • Designed for aqueous molecular weight determination.
  • Includes 10 vials, each with 1.0 g of molecular weight standards.
  • Molecular weight standards included: 100, 200, 400, 600, 1000, 1500, 4300, 7000, 13000, 22000.
  • Separation mode: GPC/GFC/SEC.
  • Detection methods: UV, RI/ELSD.
  • Formulation: Solid.
  • Storage condition: 4°C.
  • System type: LC.
  • Shelf life of 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient.
  • Tolerance: 10% (Mp).
  • Type: Calibration.
  • Application: Gel filtration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.