Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene glycol tested1KG
Catalog No. 502898472
Encompass
Poly(ethylene glycol) (PEG) is a non-toxic water-soluble polymer. PEG term is used for the polymer chains with molecular weights below 20 000 while poly(ethylene oxide) (PEO) refers to the higher molecular weight polymers. PEG has various biomedical applications. It forms complexes with cyclodextrins. Engineering of PEG hydrogel particles by mesoporous silica (MS) templating method has been reported.
Catalog No. 50-289-8472 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 812421KG
Poly(ethylene glycol) (PEG) is a non-toxic water-soluble polymer. PEG term is used for the polymer chains with molecular weights below 20 000 while poly(ethylene oxide) (PEO) refers to the higher molecular weight polymers. PEG has various biomedical applications. It forms complexes with cyclodextrins. Engineering of PEG hydrogel particles by mesoporous silica (MS) templating method has been reported.