Learn More
U.S. Pharmacopeia High Density Polyethylene, 9002-88-4, MFCD00084423, 3 strips
Catalog No. NC0215268
Encompass
Molecular formula (C2H4)x, Melting Point 527 °F (275 °C)
Catalog No. NC0215268 Supplier U.S. Pharmacopeia Supplier No. 1546707
Add to Cart
Add to Cart
Molecular formula (C2H4)x, Melting Point 527 °F (275 °C)