Learn More
Kyfora Bio Polyethylene linear MW 52000
Catalog No. 5000285309
Encompass_Preferred
Polyethylene (PE) is a semi-crystalline polymer that can be used to package solid oral dosage forms It is resistant to water vapor thus making it suitable for moisture-sensitive products
Catalog No. 50-002-85309 Supplier Kyfora Bio Supplier No. 098291
Polyethylene (PE) is a semi-crystalline polymer that can be used to package solid oral dosage forms It is resistant to water vapor thus making it suitable for moisture-sensitive products