Learn More
Waters Corp Waters Polyethylene Oxide Standard, 850,000 Da, 0.5 g
Catalog No. 50785017 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polyethylene oxide standard is an individual GPC standard with a molecular weight of 850,000 Da, supplied in a 0.5 g quantity. It is designed for molecular weight determination, verifying components in sample mixtures, and simplifying identification.
  • Molecular weight of 850,000 Da
  • Package quantity of 0.5 g
  • Used to verify a molecular weight component in a sample mixture
  • Simplifies molecular weight identification
Catalog No. 50-785-017 Supplier Waters Corp Supplier No. WAT011586
This polyethylene oxide standard is an individual GPC standard with a molecular weight of 850,000 Da, supplied in a 0.5 g quantity. It is designed for molecular weight determination, verifying components in sample mixtures, and simplifying identification.
  • Molecular weight of 850,000 Da
  • Package quantity of 0.5 g
  • Used to verify a molecular weight component in a sample mixture
  • Simplifies molecular weight identification