Learn More
Starplex™ Scientific Polyethylene Plug Caps
Catalog No. 14375440 Shop All Starplex Scientific Products
Change view
Click to view available options
Color:
Light Blue
Catalog No. Color
14-375-440 Light Blue
1 options
This item is not returnable. View return policy
Catalog No. 14-375-440 Supplier Starplex™ Scientific Supplier No. V9199
Add to Cart
Add to Cart
This item is not returnable. View return policy
For temporary vial closure
  • Fit 5 and 10mL vials
  • Perform well during specimen processing
  • Nonsterile
  • Not designed for reuse
Specifications
Color Light Blue
Material Polyethylene
Sterility Non-sterile