Learn More
International Crystal Polyethylene IR Sample Cards, 50/pk
Catalog No. NC0397298
Encompass_Preferred
PTFE and polyethylene IR sample cards provide inexpensive disposable sample substrates for qualitative analysis. The 2" x 4" cards mount vertically in the universal slide mount in any spectrophotometer. The cards have a clear aperture of 19mm and space is provided to write on the card to identify the date of the analysis, the sample and the name of the analyst.
Catalog No. NC0397298 Supplier International Crystal Supplier No. 00007366
PTFE and polyethylene IR sample cards provide inexpensive disposable sample substrates for qualitative analysis. The 2" x 4" cards mount vertically in the universal slide mount in any spectrophotometer. The cards have a clear aperture of 19mm and space is provided to write on the card to identify the date of the analysis, the sample and the name of the analyst.