Learn More
Waters Corp WATERS Screw Cap, Polyethylene, Septumless, 12 x 32 mm Vials
Catalog No. 50782195 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Polyethylene Septumless Screw Cap for 12 x 32 mm Vials seals solvents, samples, and solutions. Its universal screw cap is compatible with most vials. Recommended for single injection applications as it does not reseal, and ideal for MS applications, it offers excellent solvent resistance and chemical compatibility.
  • Cap and seal are polyethylene.
  • For 12 x 32 mm vials.
  • Packaged as 100 units.
  • Screw neck closure, white cap.
  • Septumless, for single injection and MS applications.
  • Excellent solvent resistance and chemical compatibility.
  • No tools required.
  • Leak-proof design.
  • Easily recyclable.
  • Enhanced protection, economical, durable, lightweight, flexible.
Catalog No. 50-782-195 Supplier Waters Corp Supplier No. 186004169
This Polyethylene Septumless Screw Cap for 12 x 32 mm Vials seals solvents, samples, and solutions. Its universal screw cap is compatible with most vials. Recommended for single injection applications as it does not reseal, and ideal for MS applications, it offers excellent solvent resistance and chemical compatibility.
  • Cap and seal are polyethylene.
  • For 12 x 32 mm vials.
  • Packaged as 100 units.
  • Screw neck closure, white cap.
  • Septumless, for single injection and MS applications.
  • Excellent solvent resistance and chemical compatibility.
  • No tools required.
  • Leak-proof design.
  • Easily recyclable.
  • Enhanced protection, economical, durable, lightweight, flexible.