Learn More
Kimble™ Polyethylene Snap Cap
SDPSCI_ED
Catalog No. S30864C Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
4
6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
S30864C 4
S30864E 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30864C Supplier Kimble™ Supplier No. 28150R4/EMD
Snap cap tight fit prevents leakage or evaporation.
For Kimax™ Series 28008, 28010, and 28012 volumetric flasks.
Order Info
Sold individually.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Yellow
Closure Material Polyethylene
For Use With (Equipment) 100 mL Volumetric Flask
Product Type Snap Cap
Closure Type Snap
Closure Size 4