Learn More
Electron Microscopy Sciences High-Density Polyethylene Staining Jar
SDP
Catalog No. 5026516
Encompass_Preferred
An unbreakable, high-density polyethylene Staining Jar with internal grooves and a shallow screw cap. The jar holds 5 single 1"x3" slides or 10 back-to-back slides vertically.
Catalog No. 50-265-16 Supplier Electron Microscopy Sciences Supplier No. 70319
An unbreakable, high-density polyethylene Staining Jar with internal grooves and a shallow screw cap. The jar holds 5 single 1"x3" slides or 10 back-to-back slides vertically.