Learn More
Consolidated Plastics Tote Box, 21" x 15 1/2" x 5" Polyethylene Quantity per case: 12
SDP
Catalog No. NC9060060
Encompass
Our Poly Tote Boxes are Injection molded of heavy-duty polyethylene material. Impervious to acids, fats, oil, grease. Easy carry handles. Temperature range is 20 Deg F to 190 Deg F. Color is gray. Our heavy duty plastic tote boxes comply with FDA regulations.
Catalog No. NC9060060 Supplier Consolidated Plastics Supplier No. 67009
Our Poly Tote Boxes are Injection molded of heavy-duty polyethylene material. Impervious to acids, fats, oil, grease. Easy carry handles. Temperature range is 20 Deg F to 190 Deg F. Color is gray. Our heavy duty plastic tote boxes comply with FDA regulations.