Learn More
Justrite™ Polyethylene Tray/Sump Combination For Shelf #29944 or 60 Gallon Safety Cabinet
Catalog No. 19822507 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Fits Shelf for 60 gal. Safety Cabinet
Catalog No. For Use With (Equipment)
19-822-507 Fits Shelf for 60 gal. Safety Cabinet
1 options
Catalog No. 19-822-507 Supplier Justrite™ Supplier No. 29961
Increase the efficiency and space of existing cabinetry. The Justrite™ Polyethylene Tray/Sump Combination For Shelf #29944 or 60 Gallon Safety Cabinet is a great solution to protect your cabinet from any leaks/spills with a poly tray that can fit over the shelf, or inside the bottom sump.
Specifications
Dimensions (D x W x H) 29.75 x 29.5 in.
For Use With (Equipment) Fits Shelf for 60 gal. Safety Cabinet
Material Polyethylene