Learn More
Justrite™ Polyethylene Tray/Sump Combination for Shelf #29941 or 54 Gallon Deep Slimline Safety Cabinet
Catalog No. 19822506 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Fits Shelf Model 29941 for 44 gal. Deep Slimline Cabinet
Catalog No. For Use With (Equipment)
19-822-506 Fits Shelf Model 29941 for 44 gal. Deep Slimline Cabinet
1 options
Catalog No. 19-822-506 Supplier Justrite™ Supplier No. 29907
Increase the efficiency and space of existing cabinetry. The Justrite™ Polyethylene Tray/Sump Combination for Shelf #29941 or 54 Gallon Deep Slimline Safety Cabinet is a great solution to protect your cabinet from any leaks/spills with a poly tray that can fit over the shelf, or inside the bottom sump.
Specifications
For Use With (Equipment) Fits Shelf Model 29941 for 44 gal. Deep Slimline Cabinet
Dimensions (D x W x H) 29 x 18.5 in.
Material Polyethylene