Learn More
Electron Microscopy Sciences Polyethylene Vial 2.5 ml, 12 mm diameter, 40 mm height
SDP
Supplier:  Electron Microscopy Sciences 7262120
Encompass_Preferred
Sample vials with a captive snap closure; flat bottom cylindrical vials with a friction-fit, flip-top closure which can not be lost. These vials are unbreakable and can withstand temperatures up to 90 degree C. The wall thickness is approx. 1 mm and these vials are ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
Diameter: 12 mm
Height: 40 mm
Catalog No. 50-949-127