Learn More
Polyethyleneglycol 60 Castor Oil, Hydrogenated, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| Color | Off-white solid |
| Acid Value | 1.5 |
| CAS Max % | 1 |
| Quantity | 450 lb. |
| Hydroxyl Value | 50-70 |
| Packaging | Drum |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.