Learn More
Research Products International Corp PEG 8000 [Polyethylene Glycol 8000], 1 Kilogram
SDP
Supplier:  Research Products International Corp P480801000.0
Encompass_Preferred
Ultrapure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.Melting Point: 55°-60°C
Catalog No. 50-213-630