Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethyleneimine, ∽ M.N. 60,000, 50 wt.% aq. solution, branched, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
Packaging:
Glass bottle
9002-98-6, 7732-18-5
C2H5N
43.069
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
9033
CHEBI:30969
aziridine
C1CN1
9002-98-6, 7732-18-5
C2H5N
43.069
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
9033
CHEBI:30969
aziridine
C1CN1
Density 1.0700g/mL
Linear Formula (-CH2CH2NH-)x-(CH2CH2N(CH2CH2NH2)-)y
Physical Form Viscous Liquid
Infrared Spectrum Authentic
Percent Purity 48 to 52% (solids)
Packaging Glass bottle
Solubility Solubility in water: soluble.
Specific Gravity 1.07
Boiling Point 100.0°C
Color Undesignated
Quantity 1kg
Flash Point >110°C
Chemical Name or Material Polyethyleneimine, 50 wt. % Aqueous Solution, Average M.W. 50-60000
Show More Show Less

RTECSNumber : TQ7917000

TSCA : TSCA