Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched

Catalog No. AC178572500
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC178572500 250 g Glass bottle
AC178571000 100 g Glass bottle
AC178570010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC178572500 Supplier Thermo Scientific Chemicals Supplier No. 178572500
  • $145.00
    $115.65 / Each of 1

    Save $29.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6 , 7732-18-5
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Physical Form Viscous Liquid
Boiling Point 100.0°C
Linear Formula (-CH2CH2NH-)x-(CH2CH2N(CH2CH2NH2)-)y
Density 1.0700g/mL
Solubility Information Solubility in water: soluble.
Quantity 250 g
Percent Purity 48 to 52% (solids)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.07
Chemical Name or Material Polyethyleneimine, 50 wt. % Aqueous Solution, Average M.W. 50-60000
Show More Show Less
RTECSNumber TQ7917000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.