Learn More
Kyfora Bio Polyethylenimine, Branched, MW 10,000 (bPEI10k) 100GR
Catalog No. 502559828
Encompass
Branched Polyethyleneimine, Mw 10,000 (bPEI 10000) is branched liquid water soluble polyamine with a high cationic charge density that readily binds anionic substrates. bPEI 10000 contains primary, secondary, and tertiary amine groups in approximately 25/50/25 ratio. It can be used in the formulation of nanocarriers, such as PEI coated graphene oxide nanoparticles or PEI coated mesoporous silica nanocarriers, for a combination of nucleic acid and drug delivery. Key application(s): Drug delivery, gene delivery
Catalog No. 50-255-9828 Supplier Kyfora Bio Supplier No. 19850100
Branched Polyethyleneimine, Mw 10,000 (bPEI 10000) is branched liquid water soluble polyamine with a high cationic charge density that readily binds anionic substrates. bPEI 10000 contains primary, secondary, and tertiary amine groups in approximately 25/50/25 ratio. It can be used in the formulation of nanocarriers, such as PEI coated graphene oxide nanoparticles or PEI coated mesoporous silica nanocarriers, for a combination of nucleic acid and drug delivery. Key application(s): Drug delivery, gene delivery