Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyethylenimine, 80% ethoxylated solution

Catalog No. 306185100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-618-5100G 100 g
30-618-5250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-618-5100G Supplier Sigma Aldrich Supplier No. 306185100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Additive to coatings and printing inks, brightener in electroplating, cationic modifier in clays and starches and adhesion/substantivity promoter.

Specifications

CAS 26658-46-8
Density 1.08 g/mL (at 25°C)
Quantity 100 g
MDL Number MFCD00243183
Synonym PEI
Molecular Weight (g/mol) 110000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.