Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyethylenimine, branched

Catalog No. 408719250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-871-9250ML 250 mL
40-871-91L 1 L
40-871-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 40-871-9250ML Supplier Sigma Aldrich Supplier No. 408719250ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Detergents, adhesives, water treatment, printing inks, dyes, cosmetics, and paper industry, adhesion promoter, lamination primer, fixative agent, flocculant, cationic dispersant, stability enhancer, surface activator, chelating agent, scavenger for aldehydes and oxides.

Specifications

CAS 25987-06-8
Density 1.050 g/mL (at 25°C)
Refractive Index n20/D 1.5240
Linear Formula H(NHCH2CH2)nNH2
MDL Number MFCD00084427
Quantity 250 mL
Synonym PEI; ethylenediamine branched
Molecular Weight (g/mol) 600
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.