Learn More
Kyfora Bio Polyethylenimine, Linear MW 100000 (LPEI 100K) 2 GR
Supplier:  Kyfora Bio 254142
Encompass_Preferred
Polyethylenimine, Linear, MW 100,000 (LPEI 100K) is a highly-charged linear cationic polymer that readily binds highly anionic substrates. It can be used as a flocculant to remove contaminants including RNA, DNA, and lipids from cellular homogenates before further purification downstream. Key application(s): Cell debris removal
Catalog No. 50-255-9786