Learn More
Electron Microscopy Sciences Polyfin Embedding Medium 1 KG
SDP
Catalog No. 5027986
Encompass_Preferred
Polyfin is a mixture of highly refined paraffins and co-polymer alloys in a convenient wax pellet. This unique formulation provides optimal tissue support while maintaining exceptional clarity. Our Polyfin has been institutionally tested and found to function flawlessly in pressure/vacuum fluid flow processors. Its low melting point (55 degree C) eliminates any tissue distortion caused by excessive heat during processing and has been demonstrated to have the least compression of seven popular paraffins.
Catalog No. 50-279-86 Supplier Electron Microscopy Sciences Supplier No. 1928008
Polyfin is a mixture of highly refined paraffins and co-polymer alloys in a convenient wax pellet. This unique formulation provides optimal tissue support while maintaining exceptional clarity. Our Polyfin has been institutionally tested and found to function flawlessly in pressure/vacuum fluid flow processors. Its low melting point (55 degree C) eliminates any tissue distortion caused by excessive heat during processing and has been demonstrated to have the least compression of seven popular paraffins.