Learn More
Cole-Parmer Cole-Parmer Polyfoam 6 Place Floating Racks for 15/50 mL Centrifuge Tubes; 10/Pk
Catalog No. NC1347453 Shop All Cole-Parmer Products
Encompass_Preferred
Economical polyfoam floating racks are easy to use in water baths or beakers. Ideal for use with PCR tubes, microtubes, and centrifuge tubes.
Catalog No. NC1347453 Supplier Cole-Parmer Supplier No. 0673954
Economical polyfoam floating racks are easy to use in water baths or beakers. Ideal for use with PCR tubes, microtubes, and centrifuge tubes.