Learn More
StatLab Polyform-F Granules
SDP
Catalog No. 21402149
Change view
Click to view available options
Material:
Proprietary
Catalog No. Material
21-402-149 Proprietary
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-149 Supplier StatLab Supplier No. PF04
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Aldehyde Spill Control Agent with safe and rapid handling of formaldehyde vapors
  • Spill control for safe and rapid handling of formaldehyde vapors
  • Formaldehyde is neutralized and irreversibly polymerized leaving a non-hazardous biodegradable polymer
  • 1 gallon
Specifications
Material Proprietary
Type Formalin Neutralizer