Learn More
Polysciences, Inc. PolyFreeze (Tissue Freezing Medium) - Clear
SDP
Catalog No. NC0391709
Encompass_Preferred
A support matrix or form of embedding medium for frozen sectioning. This medium freezes quickly supporting the tissue for sectioning at 4 and up with no cracking of the matrix at temperatures from -8C to -25C. Snap freeze using PolyFreeze with isopentane and liquid nitrogen or dry ice (slush/slurry or bunker). Store specimens frozen in PolyFreeze in liquid nitrogen canisters or in airtight containers in a -80C freezer.
Catalog No. NC0391709 Supplier Polysciences, Inc. Supplier No. 196364
A support matrix or form of embedding medium for frozen sectioning. This medium freezes quickly supporting the tissue for sectioning at 4 and up with no cracking of the matrix at temperatures from -8C to -25C. Snap freeze using PolyFreeze with isopentane and liquid nitrogen or dry ice (slush/slurry or bunker). Store specimens frozen in PolyFreeze in liquid nitrogen canisters or in airtight containers in a -80C freezer.