Learn More
Cayman Chemical Tenuigenin, Purity: ≥98%, CAS: 2469-34-3, 10 mg
SDP
Catalog No. 502940497
Encompass_Preferred
A triterpenoid sapogenin; inhibits IL-1β and ROS production from LPS-induced mouse BV2 microglia at 4 μM; decreases NLRP3 inflammasome activation and prevents dopaminergic neuronal degeneration in the mouse substantia nigra pars compacta in an MPTP-induced mouse model of Parkinson’s disease at 25 mg/kg per day; reduces increases in lung edema and BALF IL-1β and TNF-α levels in a mouse model of intranasal S. aureus-induced pneumonia at 2 mg/kg
Catalog No. 50-294-0497 Supplier Cayman Chemical Supplier No. 2882410 MG
A triterpenoid sapogenin; inhibits IL-1β and ROS production from LPS-induced mouse BV2 microglia at 4 μM; decreases NLRP3 inflammasome activation and prevents dopaminergic neuronal degeneration in the mouse substantia nigra pars compacta in an MPTP-induced mouse model of Parkinson’s disease at 25 mg/kg per day; reduces increases in lung edema and BALF IL-1β and TNF-α levels in a mouse model of intranasal S. aureus-induced pneumonia at 2 mg/kg