Learn More
Cayman Chemical Polygalacic Acid, Purity: ≥95%, CAS: 22338-71-2, 10 mg
SDP
Catalog No. 502991215
Encompass_Preferred
A triterpenoid saponin with antioxidant and neuroprotective activities; scavenges peroxyl and peroxynitrite radicals in TOSC assays from 25-200 µM; reverses scopolamine-induced memory deficits, decreases in hippocampal ACh levels, increases in hippocampal IL-1β and IL-10 levels, and decreases in brain SOD and GSH levels in mice at 3, 6, and 12 mg/kg
Catalog No. 50-299-1215 Supplier Cayman Chemical Supplier No. 3062010 MG
A triterpenoid saponin with antioxidant and neuroprotective activities; scavenges peroxyl and peroxynitrite radicals in TOSC assays from 25-200 µM; reverses scopolamine-induced memory deficits, decreases in hippocampal ACh levels, increases in hippocampal IL-1β and IL-10 levels, and decreases in brain SOD and GSH levels in mice at 3, 6, and 12 mg/kg