Learn More
Medchemexpress LLC HY-N6064 1mg Medchemexpress, Polygalacin D CAS:66663-91-0 Purity:>98%
Supplier: Medchemexpress LLC HYN60641MG
Medchemexpress, HY-N6064 1mg Polygalacin D CAS:66663-91-0 Polygalacin D (PGD) is a bioactive compound isolated from Platycodon grandiflorum (Jacq.) with anticancer and anti-proliferative properties.PGD suppresses the expression of the IAP family of proteins including survivin, cIAP-1 and cIAP-2 and blocks the PI3K/Akt pathway by inhibiting the phosphorylation of GSK3β, Akt and the expression of PI3K. Polygalacin D induces apoptosis Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.