Learn More
Sigma Aldrich Fine Chemicals Biosciences Polygalacturonic acid 85 t100G
Catalog No. 502902363
Encompass
Polygalacturonic acid is composed of galacturonic acid unit arranged as a long polymer chain. Methyl esters of polygalacturonic acid forms pectin.
Catalog No. 50-290-2363 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P3889100G
Polygalacturonic acid is composed of galacturonic acid unit arranged as a long polymer chain. Methyl esters of polygalacturonic acid forms pectin.