Learn More
Sigma Aldrich Fine Chemicals Biosciences Polygalacturonic acid 85 t500G
Catalog No. 502914322
Encompass
Polygalacturonic acid is composed of galacturonic acid unit arranged as a long polymer chain. Methyl esters of polygalacturonic acid forms pectin.
Catalog No. 50-291-4322 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P3889500G
Polygalacturonic acid is composed of galacturonic acid unit arranged as a long polymer chain. Methyl esters of polygalacturonic acid forms pectin.