Learn More
Cayman Chemical Polygalae radx Tenufoln 10mg
SDP
Catalog No. 502992142
Encompass_Preferred
A saponin with neuroprotective activity; inhibits BACE activity by 54.4% at 10 μg/ml; inhibits secretion of Aβ40 and Aβ42 from COS-7 cells expressing APP695 WT or APP695 Swe at 2 μg/ml; prevents decreases in PC12 cell viability induced by Aβ (25-35) at 20 and 40 μg/L; inhibits increases in the escape latency and total swimming distance induced by Aβ (25-35) in the Morris water maze in mice at 3 and 9 mg/kg
Catalog No. 50-299-2142 Supplier Cayman Chemical Supplier No. 3061710 MG
A saponin with neuroprotective activity; inhibits BACE activity by 54.4% at 10 μg/ml; inhibits secretion of Aβ40 and Aβ42 from COS-7 cells expressing APP695 WT or APP695 Swe at 2 μg/ml; prevents decreases in PC12 cell viability induced by Aβ (25-35) at 20 and 40 μg/L; inhibits increases in the escape latency and total swimming distance induced by Aβ (25-35) in the Morris water maze in mice at 3 and 9 mg/kg