Learn More
Medchemexpress LLC Polygalasaponin F | 882664-74-6 | 1091.24 | 10 MG
SDP
Catalog No. 5000389290
Encompass_Preferred
Polygalasaponin F is an orally active triterpenoid saponin monomer with broad biological activity, modulating several key cellular pathways.
  • Downregulates Bax, p53, caspase-3, NF-κB p65, and MEK1.
  • Restores and upregulates Bcl-2.
  • Activates PI3K/Akt signaling pathway.
  • Inhibits p38 MAPK phosphorylation, NF-κB translocation, TLR4-mediated signaling, mitophagy, and ROS production.
  • Enhances cell viability, suppresses apoptosis, and maintains mitochondrial function.
  • Alleviates Ca2+ overload and inhibits inflammatory cytokine release.
  • Alleviates lung injury induced by influenza A H1N1.
  • Alleviates cerebral ischemia-reperfusion injury.
  • Applicable for research in neurodegenerative diseases and infectious diseases.
Catalog No. 50-003-89290 Supplier Medchemexpress LLC Supplier No. HYN039210MM/1ML
Polygalasaponin F is an orally active triterpenoid saponin monomer with broad biological activity, modulating several key cellular pathways.
  • Downregulates Bax, p53, caspase-3, NF-κB p65, and MEK1.
  • Restores and upregulates Bcl-2.
  • Activates PI3K/Akt signaling pathway.
  • Inhibits p38 MAPK phosphorylation, NF-κB translocation, TLR4-mediated signaling, mitophagy, and ROS production.
  • Enhances cell viability, suppresses apoptosis, and maintains mitochondrial function.
  • Alleviates Ca2+ overload and inhibits inflammatory cytokine release.
  • Alleviates lung injury induced by influenza A H1N1.
  • Alleviates cerebral ischemia-reperfusion injury.
  • Applicable for research in neurodegenerative diseases and infectious diseases.