Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polygalasaponin V | 162857-65-0 | 5 MG
SDP

Catalog No. 5000420951
Encompass_Preferred

Polygalasaponin V is a triterpenoid saponin isolated from the aerial parts of *Polygala japonica*. This compound has been studied for its potential expectorant, anti-inflammatory, antibacterial, and antidepressant properties, reflecting its traditional use in folk medicine.

  • Isolated from *Polygala japonica*
  • Molecular formula: C58H94O27
  • Molecular weight: 1223.35
  • Purity: 99.94%
  • Appearance: White to off-white solid
  • Classified as a triterpenoid saponin

Catalog No. 50-004-20951 Supplier Medchemexpress LLC Supplier No. HYN21695MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polygalasaponin V is a triterpenoid saponin isolated from the aerial parts of *Polygala japonica*. This compound has been studied for its potential expectorant, anti-inflammatory, antibacterial, and antidepressant properties, reflecting its traditional use in folk medicine.

  • Isolated from *Polygala japonica*
  • Molecular formula: C58H94O27
  • Molecular weight: 1223.35
  • Purity: 99.94%
  • Appearance: White to off-white solid
  • Classified as a triterpenoid saponin

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.