Learn More
Medchemexpress LLC Polygalasaponin V | 162857-65-0 | MFCD13186870 | 99.9% | 1223.35 | C58H94O27 | 10 MG
SDP
Catalog No. 5000296364
Encompass_Preferred
Polygalasaponin V is a triterpenoid saponin isolated from the aerial parts of Polygala japonica and provided as a research-grade natural product. It is intended for biochemical and pharmacological studies and supplied as a high-purity solid in small pack sizes for laboratory use.
  • High purity suitable for analytical and biological assays.
  • Available in small research pack sizes for convenient testing.
  • Stable when stored protected from light at recommended temperatures.
  • Molecular weight 1223.35 and formula C58H94O27 provided for experimental reference.
  • Supplied with documentation such as a datasheet for handling and storage information.
Catalog No. 50-002-96364 Supplier Medchemexpress LLC Supplier No. HYN216910MG
Polygalasaponin V is a triterpenoid saponin isolated from the aerial parts of Polygala japonica and provided as a research-grade natural product. It is intended for biochemical and pharmacological studies and supplied as a high-purity solid in small pack sizes for laboratory use.
  • High purity suitable for analytical and biological assays.
  • Available in small research pack sizes for convenient testing.
  • Stable when stored protected from light at recommended temperatures.
  • Molecular weight 1223.35 and formula C58H94O27 provided for experimental reference.
  • Supplied with documentation such as a datasheet for handling and storage information.