Learn More
Medchemexpress LLC Polygalaxanthone III | 162857-78-5 | 568.48 | 1 ML
SDP
Catalog No. 5000416159
Encompass_Preferred
Polygalaxanthone III is extracted from *polygala tenuifolia wild* and has an inhibitory effect on the CYP450 enzyme. It inhibits chlorzoxazone 6-hydroxylation catalyzed by CYP2E1 with an IC50 of 50.56 μM.
  • Inhibitory effect towards CYP450 enzyme
  • Specifically inhibits chlorzoxazone 6-hydroxylation catalyzed by CYP2E1
Catalog No. 50-004-16159 Supplier Medchemexpress LLC Supplier No. HYN140710MM/1ML
Polygalaxanthone III is extracted from *polygala tenuifolia wild* and has an inhibitory effect on the CYP450 enzyme. It inhibits chlorzoxazone 6-hydroxylation catalyzed by CYP2E1 with an IC50 of 50.56 μM.
  • Inhibitory effect towards CYP450 enzyme
  • Specifically inhibits chlorzoxazone 6-hydroxylation catalyzed by CYP2E1