Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polygalaxanthone XI | 857859-82-6 | 5 MG
SDP

Catalog No. 5000419407
Encompass_Preferred

Polygalaxanthone XI is a xanthone glycoside isolated from the cortexes of Polygala tenuifolia. It can be utilized in the study of expectorant and tranquilizing agents. It is intended for research use only and is not sold to patients.

  • Isolated from the cortexes of Polygala tenuifolia
  • Can be used in the study of expectorant and tranquilizing agents
  • For research use only
  • Molecular weight: 568.48
  • Chemical formula: C25H28O15
  • Appearance: solid, light yellow to yellow color
  • Purity: 99.0%

Catalog No. 50-004-19407 Supplier Medchemexpress LLC Supplier No. HYN68035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polygalaxanthone XI is a xanthone glycoside isolated from the cortexes of Polygala tenuifolia. It can be utilized in the study of expectorant and tranquilizing agents. It is intended for research use only and is not sold to patients.

  • Isolated from the cortexes of Polygala tenuifolia
  • Can be used in the study of expectorant and tranquilizing agents
  • For research use only
  • Molecular weight: 568.48
  • Chemical formula: C25H28O15
  • Appearance: solid, light yellow to yellow color
  • Purity: 99.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.