Learn More
Cayman Chemical Polygalaxnthon III 10mg
SDP
Catalog No. 502988231
Encompass_Preferred
A xanthone glycoside with antioxidant activity; scavenges hydroxyl and DPPH radicals in cell-free assays (IC50s = 83.5 and 76.1 µM, respectively)
Catalog No. 50-298-8231 Supplier Cayman Chemical Supplier No. 3062110 MG
A xanthone glycoside with antioxidant activity; scavenges hydroxyl and DPPH radicals in cell-free assays (IC50s = 83.5 and 76.1 µM, respectively)