Learn More
Medchemexpress LLC HY-N1479 5mg Medchemexpress, Polygalic acid CAS:1260-04-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14795MG
Encompass_Preferred
Medchemexpress, HY-N1479 5mg Polygalic acid CAS:1260-04-4 Polygalic acid, a triterpenoid saponin, is considered one of the major active constituents of Polygala tenuifolia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1449